missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Gastrokine 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6395
This item is not returnable.
View return policy
Description
Gastrokine 1 Monoclonal specifically detects Gastrokine 1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| Gastrokine 1 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q9NS71 | |
| GKN1 | |
| E. coli-derived recombinant human Gastrokine 1 Asn35-Asn199 Accession # Q9NS71 | |
| 100 μg | |
| Primary | |
| Detects human Gastrokine 1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Gastrokine 1 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 665701 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 18 kDa antrum mucosa protein, AMP18, AMP-18, AMP18BRICHOS domain containing 1, BRICD1, CA11, CA11MGC70354, FOV, foveolin, gastrokine 1, gastrokine-1, GKN1, Protein CA11 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 56287 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction