missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GASP-2/WFIKKN Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2136
This item is not returnable.
View return policy
Description
GASP-2/WFIKKN Monoclonal specifically detects GASP-2/WFIKKN in Human samples. It is validated for Western Blot.
Specifications
| GASP-2/WFIKKN | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q96NZ8 | |
| WFIKKN1 | |
| Mouse myeloma cell line NS0-derived recombinant human GASP-2/WFIKKN Ala20-Asp548 Accession # Q96NZ8 | |
| 100 μg | |
| Primary | |
| Detects human GASP-2/WFIKKN in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human GASP-1 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 289819 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C16orf12, Chromosome 16 Open Reading Frame 12, GASP2, GASP-2, Growth And Differentiation Factor-Associated Serum Protein 2, hGASP-2, RJD2, WAP Four-Disulfide Core Domain 20A, WAP, Follistatin/Kazal, Immunoglobulin, Kunitz And Netrin Domain Containing 1, WAP, FS, Ig, KU, And NTR-Containing Protein, WFDC20A, WFIKKN, WFIKKN1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 117166 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction