missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GASP-2/WFIKKN Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB21361-SP
This item is not returnable.
View return policy
Description
GASP-2/WFIKKN Monoclonal specifically detects GASP-2/WFIKKN in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| GASP-2/WFIKKN | |
| Monoclonal | |
| LYOPH | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q96NZ8 | |
| WFIKKN1 | |
| Mouse myeloma cell line NS0-derived recombinant human GASP-2/WFIKKN Ala20-Asp548 Accession # Q96NZ8 | |
| 25 μg | |
| Primary | |
| Detects human GASP-2/WFIKKN in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human GASP-1 is observed. | |
| Human | |
| Purified |
| ELISA | |
| 289827 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C16orf12, Chromosome 16 Open Reading Frame 12, GASP2, GASP-2, Growth And Differentiation Factor-Associated Serum Protein 2, hGASP-2, RJD2, WAP Four-Disulfide Core Domain 20A, WAP, Follistatin/Kazal, Immunoglobulin, Kunitz And Netrin Domain Containing 1, WAP, FS, Ig, KU, And NTR-Containing Protein, WFDC20A, WFIKKN, WFIKKN1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 117166 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction