missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Gas6 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 14 publications
Brand: R&D Systems AF885-SP
This item is not returnable.
View return policy
Description
Gas6 Polyclonal specifically detects Gas6 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Gas6 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AXLLG, AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2621 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| NP_000811 | |
| GAS6 | |
| Mouse myeloma cell line NS0-derived recombinant human Gas6 Asp118-Ala678 Accession # NP_000811 | |
| 25 μg | |
| Primary | |
| Detects human Gas6 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant mouse Gas6 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction