missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Gas1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2636
This item is not returnable.
View return policy
Description
Gas1 Polyclonal specifically detects Gas1 in Human samples. It is validated for Western Blot.
Specifications
| Gas1 | |
| Polyclonal | |
| Unconjugated | |
| P54826 | |
| GAS1 | |
| Mouse myeloma cell line NS0-derived recombinant human Gas1 Leu40-Ser318 Accession # P54826 | |
| 100 μg | |
| Primary | |
| Detects human Gas1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse Gas1 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| GAS-1, growth arrest-specific 1, Growth arrest-specific gene-1, growth arrest-specific protein 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2619 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction