missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Human GAPD (GAPDH) Endogenous Control (VIC™/TAMRA™ probe, primer limited)
Shop All Applied Biosystems Products

Description
The Applied Biosystems™ Human GAPD (GAPDH) Endogenous Control (VIC™ ⁄ TAMRA Probe, Primer Limited) is intended as an endogenous control. It allows relative gene expression quantification in cDNA samples when used with other TaqMan™ gene expression assays. Probe is labeled with VIC™ dye - TAMRA™ dye and the primers are limited. Can be used for multiplex or singleplex PCR reactions. Endogenous control is to be used with Inventoried and Made to Order TaqMan™ Gene Expression Assays, Custom TaqMan™ Gene Expression Assays, and⁄or Custom TaqMan™ Primers and Probes.
Assay Details:
Gene Symbol: GAPDH
RefSeq: NM_002046.3
Probe Exon Location: 3
Amplicon Size: 118
TaqMan™ Endogenous Controls
Eliminate months of assay design, formulation, and testing by using TaqMan™ Endogenous Controls as your controls to quantify gene expression. This convenient collection of pre-designed probe and primer sets enables you to normalize the amount of sample RNA or DNA in a reaction.
Complete Solution for Quantitative Gene Expression
Having a hard time deciding what controls to use to quantify gene expression — even with detailed information on biological systems? Now, with TaqMan™ Endogenous Controls, you can avoid all the trial-and-error of selecting controls for most common human, mouse, rat, and eukaryotic genes.
Simple to Use
All components of the TaqMan™ Endogenous Controls are QC tested, formulated into a single 20X mix, and functionally tested. The controls are not only simple to use, but they are also fully compatible with universal conditions for two-step RT-PCR. Just add TaqMan™ Universal PCR Master Mix (with or without AmpErase™ UNG) and your cDNA sample to generate sensitive, reproducible, and truly quantitative gene expression data on Applied Biosystems instruments including the Applied Biosystems 7900HT, 7300, 7500 Real-Time PCR Systems, and the 7000 and 7700 Sequence Detection Systems.
Flexible Offering
We build each endogenous control using our proven 5' nuclease chemistry. For maximum flexibility, you can choose between two different reporter dyes and two quenchers:
A FAM™ dye-labeled TaqMan™ MGB probe (250nM, final concentration) and two unlabeled PCR primers (900nM each)
A VIC™ dye-labeled TaqMan™ MGB probe (250nM, final concentration) and two unlabeled PCR primers (150nM each y primer limited)
A VIC™ dye-labeled TAMRA™ probe (250nM, final concentration) and two unlabeled PCR primers (150nM each y primer limited)
Choosing the Right Endogenous Control
Endogenous controls can normalize the expression levels of target genes by correcting differences in the amount of cDNA that is loaded into PCR reaction wells. For best results, verify that the endogenous control is consistently expressed in the sample set to be tested. Endogenous control expression must be uniform across all samples in the study. For multiplexing, ensure that the gene expression level of the endogenous control is greater than that of the target.
Multiplex vs. Singleplex PCR
All TaqMan™ Endogenous Controls that contain probes labeled with the VIC™ reporter dye are primer limited. This allows multiplexing of TaqMan™ Endogenous Controls with target gene expression assays, provided that the control gene is more abundantly expressed than the target gene. All TaqMan™ Endogenous Controls that contain probes labeled with the FAM™ reporter dye are not primer limited and are not intended for multiplexing .
For Research Use Only. Not for use in diagnostic procedures.
Assay Details:
Gene Symbol: GAPDH
RefSeq: NM_002046.3
Probe Exon Location: 3
Amplicon Size: 118
TaqMan™ Endogenous Controls
Eliminate months of assay design, formulation, and testing by using TaqMan™ Endogenous Controls as your controls to quantify gene expression. This convenient collection of pre-designed probe and primer sets enables you to normalize the amount of sample RNA or DNA in a reaction.
Complete Solution for Quantitative Gene Expression
Having a hard time deciding what controls to use to quantify gene expression — even with detailed information on biological systems? Now, with TaqMan™ Endogenous Controls, you can avoid all the trial-and-error of selecting controls for most common human, mouse, rat, and eukaryotic genes.
Simple to Use
All components of the TaqMan™ Endogenous Controls are QC tested, formulated into a single 20X mix, and functionally tested. The controls are not only simple to use, but they are also fully compatible with universal conditions for two-step RT-PCR. Just add TaqMan™ Universal PCR Master Mix (with or without AmpErase™ UNG) and your cDNA sample to generate sensitive, reproducible, and truly quantitative gene expression data on Applied Biosystems instruments including the Applied Biosystems 7900HT, 7300, 7500 Real-Time PCR Systems, and the 7000 and 7700 Sequence Detection Systems.
Flexible Offering
We build each endogenous control using our proven 5' nuclease chemistry. For maximum flexibility, you can choose between two different reporter dyes and two quenchers:
Choosing the Right Endogenous Control
Endogenous controls can normalize the expression levels of target genes by correcting differences in the amount of cDNA that is loaded into PCR reaction wells. For best results, verify that the endogenous control is consistently expressed in the sample set to be tested. Endogenous control expression must be uniform across all samples in the study. For multiplexing, ensure that the gene expression level of the endogenous control is greater than that of the target.
Multiplex vs. Singleplex PCR
All TaqMan™ Endogenous Controls that contain probes labeled with the VIC™ reporter dye are primer limited. This allows multiplexing of TaqMan™ Endogenous Controls with target gene expression assays, provided that the control gene is more abundantly expressed than the target gene. All TaqMan™ Endogenous Controls that contain probes labeled with the FAM™ reporter dye are not primer limited and are not intended for multiplexing .
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
| Concentration | 20X |
| Accession Number | NM_002046.3 |
| Amplicon Size | 118 |
| Content And Storage | Control Primer-Probe Set(s) |
| Detection Method | Primer-probe |
| Form | Liquid |
| Format | Tube |
| PCR Method | 2-step RT-qPCR |
| Gene Symbol | GAPDH |
| Label or Dye | VIC |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?