missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human gamma-Synuclein Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5745-SP
This item is not returnable.
View return policy
Description
gamma-Synuclein Monoclonal specifically detects gamma-Synuclein in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| gamma-Synuclein | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q6FHG5 | |
| SNCG | |
| E. coli-derived recombinant human gamma-Synuclein Asn47-Glu120 Accession # Q6FHG5 | |
| 25 μg | |
| Primary | |
| Detects human gamma-Synuclein in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 514304 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BCSG1, BCSG1PERSYN, Breast cancer-specific gene 1 protein, gamma-synuclein, persyn, PRSN, SNCG, SRPRSN, synoretin, synuclein, gamma (breast cancer-specific protein 1), Synuclein-gamma | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6623 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction