missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GALNTL4 (aa 465-602) Control Fragment Recombinant Protein

Product Code. 30201526
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201526

Brand: Invitrogen™ RP90092

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53017 (PA5-53017. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Catalyzes the initial reaction in O-linked oligosaccharide biosynthesis, the transfer of an N-acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6P9A2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 374378
Name Human GALNTL4 (aa 465-602) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2900011G21Rik; BC024988; GalNAc-T15; GALNACT18; GalNAc-T18; GalNAc-T-like protein 4; GalNAc-transferase 18; GALNT15; GALNT18; Galntl4; LOW QUALITY PROTEIN: polypeptide N-acetylgalactosaminyltransferase 18; mpp-GalNAc-T18(O18); polypeptide GalNAc transferase 18; Polypeptide GalNAc transferase-like protein 4; polypeptide N-acetylgalactosaminyltransferase 18; polypeptide N-acetylgalactosaminyltransferase-like 4; polypeptide N-acetylgalactosaminyltransferase-like protein 4; pp-GaNTase-like protein 4; protein-UDP acetylgalactosaminyltransferase-like protein 4; putative polypeptide N-acetylgalactosaminyltransferase-like protein 4; RGD1309623; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like protein 4; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 15; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 18; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase-like 4
Common Name GALNTL4
Gene Symbol GALNT18
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SDIIAYGVLQNSLKTDLCLDQGPDTENVPIMYICHGMTPQNVYYTSSQQIHVGILSPTVDDDDNRCLVDVNSRPRLIECSYAKAKRMKLHWQFSQGGPIQNRKSKRCLELQENSDLEFGFQLVLQKCSGQHWSITNVL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.