missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human GALM (aa 54-139) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84671 (PA5-84671. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes an enzyme that catalyzes the epimerization of hexose sugars such as glucose and galactose. The encoded protein is expressed in the cytoplasm and has a preference for galactose. The encoded protein may be required for normal galactose metabolism by maintaining the equilibrium of alpha and beta anomers of galactose.
Specifications
Specifications
| Accession Number | Q96C23 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 130589 |
| Name | Human GALM (aa 54-139) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | A530057M15Rik; aldose 1-epimerase; aldose mutarotase; AU015645; AU020959; BLOCK25; epididymis secretory sperm binding protein Li 63 p; galactomutarotase; galactose enzyme activator; galactose mutarotase; galactose mutarotase (aldose 1-epimerase); GALM; GLAT; HEL-S-63 p; IBD1 |
| Common Name | GALM |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction