missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Gallbladder Membrane Tissue Lysate (Adult Membrane Tumor)

Product Code. 18166949 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18166949

Brand: Novus Biologicals™ NBL126422

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The Human Gallbladder Membrane Tissue Lysate (Adult Membrane Tumor) is derived from human.
TRUSTED_SUSTAINABILITY

Specifications

Tissue Digestive/Waste
Quantity 0.1 mg
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Membrane Protein - Human Tumor Tissue: Gallbladder
Cellular Fraction Membrane
Tissue Condition Tumor
Cross Reactivity Human
Host Species Human
Product Type Human Gallbladder Membrane Tissue Lysate (Adult Tumor)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.