missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Galectin-9 Quantikine ELISA Kit

Product Code. 18732094
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18732094

Brand: R&D Systems™ DGAL90

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Galectin-9 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Galectin-9 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Galectin-9 in cell culture supernates (100 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul), saliva (100 ul), urine (10 ul), human milk (10 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.028 ng/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Specificity Natural and Recombinant Human Galectin-9
For Use With (Application) ELISA
Gene ID (Entrez) 3965
Gene Symbol LGALS9
Quantity 1 Kit
Sample Volume Cell Culture Supernates (100uL),Serum (25uL),EDTA Plasma (25uL),Heparin Plasma (25uL),Saliva (100uL),Urine (10uL),Human Milk (10uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym ecalectin, gal-9, GAL9, galectin 9, galectin-9, Galectin9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein
Target Galectin-9
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.