missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Galectin-9 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£177.00 - £440.00
Specifications
| Antigen | Galectin-9 |
|---|---|
| Clone | 2315B |
| Dilution | 0.25 μg/106 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18705765
![]() |
R&D Systems
MAB9064-100 |
100 μg |
£440.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18795815
![]() |
R&D Systems
MAB9064-SP |
25 μg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Human samples. It is validated for Flow CytometrySpecifications
| Galectin-9 | |
| 0.25 μg/106 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| ecalectin, Gal-9, galectin 9, galectin-9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein | |
| Human embryonic kidney cell, HEK293 derived human Galectin-9, Met1-Thr323, Accession # O00182 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2315B | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O00182 | |
| 3965 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title