missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Galectin-7 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF1339-SP
This item is not returnable.
View return policy
Description
Galectin-7 Polyclonal specifically detects Galectin-7 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| Galectin-7 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Gal-7, GAL7, galectin 7, galectin-7, Galectin7, HKL-14, lectin, galactoside-binding, soluble, 7, LGALS7, LGALS7A, p53-induced gene 1 protein, PI7, PIG1, TP53I1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3963 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, ELISA | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| NP_002298 | |
| LGALS7 | |
| E. coli-derived recombinant human Galectin-7 Ser2-Phe136 Accession # NP_002298 | |
| 25 μg | |
| Primary | |
| Detects human Galectin-7 in ELISAs and Western blots. In sandwich immunoassays, approximately 50% cross-reactivity with recombinant mouse (rm) Galectin-7 is observed and less than 0.5% cross-reactivity with recombinant human (rh) Galectin-1, rmGalectin-3, rhGalectin-4, and rhGalectin-8 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction