missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Galectin-3BP/MAC-2BP Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
The Human Galectin-3BP/MAC-2BP Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Galectin-3BP/MAC-2BP/LGALS3BP.
Specifications
Specifications
| Assay Sensitivity | 0.161 ng/mL |
| Sample Type | Cell Culture Supernatant, Saliva |
| Quantity | 1 Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction