missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Galectin-3 Quantikine ELISA Kit
Click to view available options
Quantity:
1 Pk. (6 Plates)
Unit Size:
Pack of 96
Description
The Human Galectin-3 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Galectin-3 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
Specifications
Specifications
| Assay Range | 0.31 to 10 ng/mL |
| Assay Sensitivity | 0.085 ng/mL |
| Conjugate | HRP |
| Cross Reactivity | < 0.5% cross-reactivity observed with available related molecules, Cross-species reactivity observed with 1 or more species tested |
| Immunoassay Kit Format | 96-well Strip Plate |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (EDTA, Heparin), Plasma (Heparin), Serum |
| Specificity | Natural and Recombinant Human Galectin-3. This assay cross-reacts 58% with Mouse Galectin-3. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3958 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction