missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ Human Galectin-3 ELISA Kit
Shop All Proteintech Products
Click to view available options
Target Species:
Human
Unit Size:
96 tests
Description
- Recognizes human Galectin-3
- Galectins are a family of animal lectins defined by shared characteristic amino acid sequences and affinity for β-galactose-containing oligosaccharides
- Galectin-3 is a 31 kDa member of β-galactoside-binding proteins with one carbohydrate recognition domain and a proline and glycine rich N-terminal domain through which is able to form oligomers
- Widely expressed in many normal tissues and tumors
- Found intracellularly in nucleus and cytoplasm or secreted outside of cell, present on cell surface or in extracellular space
- Involved in biological processes (cell growth, adhesion, differentiation, apopotosis, angiogenesis, immune response, neoplastic transformation, metastasis)
- Elevated serum Galectin-3 levels reported in patients with breast, gastrointestinal, lung or ovarian cancer, melanoma and non-Hodgkin's lymphoma
Process:
- Antibody specific for human Galectin-3 pre-coated onto microwells
- Human Galectin-3 protein in samples captured by the coated antibody after incubation
- After extensive washing, another antibody of biotinylated specific for human Galectin-3 is added to detect captured human Galectin-3 protein
- For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent
- Solution containing sulfuric acid used to stop color development and color intensity
- Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm
Specifications
Specifications
| Accession Number | BC001120 |
| Assay Range | 0.625 to 40 ng/mL |
| Assay Sensitivity | 0.16 ng/mL |
| Certifications/Compliance | ISO 13485 |
| Detection Method | Sandwich ELISA, Double Antibody |
| Immunoassay Kit Format | Coated ELISA Kit |
| Sample Type | Plasma, Serum |
| Substrate | DAB |
| Target Species | Human |
| Gene ID (Entrez) | 3958 |
| Show More |
Safety and Handling
Product Identifier
- 96TEST Human TNF-alpha ELISA Kit
- Warning
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P234-Keep only in original container.
- P264-Wash skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction