missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Galectin-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB11541-500
This item is not returnable.
View return policy
Description
Galectin-3 Monoclonal specifically detects Galectin-3 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| Galectin-3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 1-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P17931.5 | |
| LGALS3 | |
| E. coli-derived recombinant human Galectin-3 Ala2-Ile250 Accession # P17931.5 | |
| 500 μg | |
| Primary | |
| Detects human Galectin-3 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human Galectin-1, 2, 4, 7, 8, or 10 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, ELISA | |
| 194805 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AGE-R3, Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, CBP35, Gal-3, GAL3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, Galectin3, GALIG, IgE-binding protein, L29, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, LGALS3, Mac-2, Mac-2 antigen, MAC2GAL3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3958 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction