missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Galectin-10 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5447-SP
This item is not returnable.
View return policy
Description
Galectin-10 Polyclonal specifically detects Galectin-10 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| Galectin-10 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Charcot-Leyden crystal proteinLGALS10A, CLC, EC 3.1.1.5, eosinophil lysophospholipase, Gal-10, GAL10, galectin 10, galectin-10, Galectin10, LGALS10, LPPL_HUMAN, Lysolecithin acylhydrolase, MGC149659 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1178 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q05315 | |
| CLC | |
| E. coli-derived recombinant human Galectin-10 Ser2-Arg142 Accession # Q05315 | |
| 25 μg | |
| Primary | |
| Detects human Galectin-10 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human Galectin-1, -2, -3, -4, -7, -8, -9 and -14 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction