missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Galectin-1 Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP8622
Code : LE
Additional Details : Weight : 0.01000kg
Description
Galectin 1 (human) Recombinant Protein, RP-8622, from Invitrogen.
Galectin-1 belongs to a large family of carbohydrate-binding proteins called lectins. Galectin-1can be either monomeric or homodimeric and is found in a wide variety of cells and tissue types. Galectin-1 can control cell growth, proliferation, induce apoptosis of activated T cells while it can also modulate cytokine secretion or inhibit pro-inflammatory cytokine production. Galectin-1 plays an important role in acute and chronic inflammation. Recombinant human Galectin-1 is a non-glycosylated protein, containing134 amino acids, with a molecular weight of 14.5 kDa. Purity is typically greater than 95% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the ability to induce chemotaxis of human blood monocytes or THP-1 with a typical range of 0.5-3.0 ug/mL. The specific activity of this protein is 2x10^3 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: ACGLVASNLN LKPGECLRVR GEVAPDAKSF VLNLGKDSNN LCLHFNPRFN AHGDANTIVC NSKDGGAWGT EQREAVFPFQ PGSVAEVCIT FDQANLTVKL PDGYEFKFPN RLNLEAINYM AADGDFKIKC VAFD.Specifications
P09382 | |
Control | |
3956 | |
Human Galectin 1 | |
50μg | |
-20°C, Avoid Freeze/Thaw Cycles | |
H-Galectin-1 | |
LGALS1 | |
Unconjugated | |
E. coli | |
Lyophilized | |
purified |
0.1mg/mL | |
10mM sodium phosphate with no preservative; pH 7.5 | |
∽14.5 | |
7.5 | |
E. coli | |
RUO | |
Human Galectin 1 | |
Human | |
Hu | |
Recombinant | |
Recombinant |
in vitro research only.