missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Galectin-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 11 publications
Brand: R&D Systems AF1152-SP
This item is not returnable.
View return policy
Description
Galectin-1 Polyclonal specifically detects Galectin-1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| Galectin-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
| 14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, BHL, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, Galectin1, GBP, HBL, HLBP14, HPL, L-14, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, LGALS1, Putative MAPK-activating protein PM12, S-Lac lectin 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3956 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Store the unopened product at -20°C to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P09382 | |
| LGALS1 | |
| E. coli-derived recombinant human Galectin-1 Ala2-Asp135 Accession # P09382 | |
| 25 μg | |
| Primary | |
| Detects human Galectin-1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction