missing translation for 'onlineSavingsMsg'
Learn More

Human Galanin R1/GALR1 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£167.00 - £412.00

Specifications

Antigen Galanin R1/GALR1
Clone 613207
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Applications Flow Cytometry, CyTOF
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18791427
GREENER_CHOICE
View Documents
R&D Systems
MAB10353-SP
25 μg
£167.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18700717
GREENER_CHOICE
View Documents
R&D Systems
MAB10353-100
100 μg
£412.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Galanin R1/GALR1 Monoclonal specifically detects Galanin R1/GALR1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications

Specifications

Galanin R1/GALR1
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Monoclonal
Purified
RUO
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
GAL1, galanin receptor 1, galanin receptor type 1, GalaninR1, GALNR1, GALNRGALNR1GAL1-R, GALR1, GALR-1
NS0 mouse myeloma cell line transfected with human Galanin R1/GALR1, Accession # P47211
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Galanin R1/GALR1 in direct ELISAs.
613207
Flow Cytometry, CyTOF
Unconjugated
Mouse
Human
P47211
2587
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.