missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GAK Antibody, R&D Systems™

Description
GAK Monoclonal specifically detects GAK in Human samples. It is validated for Western Blot.
Tekniske data
Tekniske data
| Antigen | GAK |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 706306 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | O14976 |
| Gene Alias | Auxillin 2, cyclin G associated kinase, cyclin-G-associated kinase, EC 2.7.11.1, FLJ16629, FLJ40395, MGC99654 |
| Vis mere |
For Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?