Learn More
Invitrogen™ Human GADD45GIP1 (aa 84-117) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-111409 (PA5-111409, PA5-64199 (PA5-64199. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q8TAE8 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 90480 |
| Name | Human GADD45GIP1 (aa 84-117) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 2310040G17Rik; 39 S ribosomal protein L59, mitochondrial; AI425883; CKBBP2; CKbetaBP2; CKII beta binding protein 2; CKII beta-associating protein; CR6 interacting factor 1; CR6-interacting factor 1; CRIF1; GADD45G interacting protein 1; GADD45GIP1; growth arrest- and DNA damage-inducible GADD45G-interacting protein; Growth arrest and DNA damage-inducible proteins-interacting protein 1; growth arrest and DNA-damage-inducible proteins-interacting protein 1; growth arrest and DNA-damage-inducible, gamma interacting protein 1; Mitochondrial large ribosomal subunit protein mL64; Mrpl59; MRP-L59; p53-responsive gene 6 protein; papillomavirus L2 interacting nuclear protein 1; Papillomavirus L2-interacting nuclear protein 1; PLINP; PLINP1; PLINP-1; PRG6 |
| Common Name | GADD45GIP1 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.