missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human GABPB1 (aa 161-255) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54045 (PA5-54045. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Transcription factor capable of interacting with purine rich repeats (GA repeats). Necessary for the expression of the Adenovirus E4 gene.
Spezifikation
Spezifikation
| Accession Number | Q06547 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 2553 |
| Name | Human GABPB1 (aa 161-255) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | BABPB2; E4TF1; E4TF1-47; E4TF1-53; E4Tf1B; GA binding protein subunit beta1; GA binding protein transcription factor beta subunit 1; GA binding protein transcription factor beta subunit 1 transcript variant gamma-2; GA binding protein transcription factor, beta subunit 1; GA repeat binding protein, beta 1; GA-binding protein subunit beta-1; Gabp beta1; GABP subunit beta-1; GABP subunit beta-2; GABPB; Gabpb1; GABPB-1; GABPB1-1; GABPB1-2; GABPB2; GABPB-2; NRF2B1; NRF2B2; Nuclear respiratory factor 2; RGD1560391; transcription factor E4TF1-47; Transcription factor E4TF1-53 |
| Common Name | GABPB1 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur