missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human G2E3 (aa 572-692) Control Fragment Recombinant Protein

Product Code. 30196218
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30196218

Brand: Invitrogen™ RP88865

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51610 (PA5-51610. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. Essential in early embryonic development to prevent apoptotic death.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q7L622
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55632
Name Human G2E3 (aa 572-692) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6030408C04Rik; 9030416F18; AW046379; D930034K21Rik; G2/M phase-specific E3 ubiquitin-protein ligase; G2/M phase-specific HECT-type E3 ubiquitin transferase; G2/M-phase specific E3 ubiquitin ligase; G2/M-phase specific E3 ubiquitin protein ligase; G2e3; hypothetical protein LOC508575; KIAA1333; mKIAA1333; PHD finger protein 7 B; PHF7B; RGD1310263
Common Name G2E3
Gene Symbol G2E3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IQAYPEAFCSILCHKPESLSAKILSELFTVHTLPDVKALGFWNSYLQAVEDGKSTTTMEDILIFATGCSSIPPAGFKPTPSIECLHVDFPVGNKCNNCLAIPITNTYKEFQENMDFTIRNT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.