Learn More
Gibco™ Human G-CSF Recombinant Protein
Description
Carrier-Free Reconstitution: We recommend that the vial be briefly centrifuged prior to opening to bring the contents to the bottom. Lyophilized hG-CSF should be reconstituted in sterile deionized water to 0.1-1.0 mg/mL to regain full activity. These stock solutions should be apportioned into working aliquots and stored at ≤-20°C. Further dilutions should be made in low endotoxin medium or buffered solution with FBS or tissue culture grade BSA. Storage: Lyophilized Hg-CSF should be stored at 2°C to 8°C, preferably desiccated. Store reconstituted hG-CSF at ≤-20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.
Specifications
Specifications
| Accession Number | P09919 |
| For Use With (Application) | Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 1440 |
| Molecular Weight (g/mol) | 19 kDa |
| Name | Human G-CSF |
| Purification Method | Purified |
| Quantity | 100 μg |
| Storage Requirements | 4°C |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.