Learn More
Gibco™ Human G-CSF Recombinant Protein
Recombinant Human Granulocyte Colony-Stimulating Factor (G-CSF) is a bioactive protein intended for use in cell culture applications
Brand: Gibco™ PHC2033
Code : 50
Additional Details : Weight : 0.50000kg
Description
Human G-CSF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
G-CSF stimulates the proliferation and differentiation of hematopoietic progenitor cells.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of mouse NFS-60 cells
- Proven Compatibility: Gibco proteins are bioassayed with Gibco media
Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Cell Culture | |
19 kDa | |
1mg | |
Human | |
Carrier-Free | |
1 mg | |
E. coli | |
Recombinant, Ligand, Mutant | |
GM-CSF, G-CSF, & M-CSF | |
Gibco™ |
1440 | |
Sequential Chromatography | |
G-CSF | |
Lyophilized hG-CSF should be stored at 2-8°C, preferably desiccated. Store reconstituted hG-CSF at ≤ -20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum. | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Signal Transduction, Oncology, Stem Cell Research, Differentiation, Drug Development | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.