missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Human G-CSF ELISA Kit

Product Code. 16841516 Shop All Proteintech Products
Change view
Click to view available options
Target Species:
Human
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Target Species Unit Size
16841516 Human 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16841516 Supplier Proteintech™ Supplier No. KE0010696T
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Solid phase sandwich ELISA used to detect and quantify protein levels of endogenous G-CSF

  • Recognizes human G-CSF
  • Granulocyte colony-stimulating factor (G-CSF, CSF3) is a protective cytokine with anti-inflammatory effects
    • Promotes survival of granulocytic lineage cells and proliferation and migration of neutrophils and trophoblast cells
    • Acts by binding to its receptor (G-CSFR) after binding with G-CSF activates canonical Janus kinase/signal transducer, activator of transcription and Ras/Raf/MAP kinase pathways
    • Potently stimulates proliferation and release of peripheral blood progenitor cells into bloodstream and is used to treat neutropenia after chemotherapy
    • Elevated levels upon intensive exercise lead to increased neutrophil counts

Process:

  • Antibody specific for G-CSF pre-coated onto microwells
  • G-CSF protein in samples captured by the coated antibody after incubation
  • After extensive washing, another antibody specific for G-CSF is added to detect captured G-CSF protein
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent
  • Solution containing sulfuric acid used to stop color development and color intensity
    • Proportional to quantity of bound protein, measurable at 450 nm with correctional wavelength set at 630 nm

Specifications

Accession Number BC033245
Assay Range 31.25 to 2000 pg/mL
Assay Sensitivity 0.4 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human
Gene ID (Entrez) 1440
Host Species Human
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 4.5 hr.
UniProt ID P09919
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human G-CSF ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.