missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FXR1 (aa 552-620) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
FXR1 is an RNA binding protein that interacts with the functionally-similar proteins FMR1 and FXR2. It may regulate intracellular transport and local transltion of certain mRNAs. These proteins shuttle between the nucleus and cytoplasm and associate with polyribosomes, predominantly with the 60S ribosomal subunit. FXR1 is required for embryonic and postnatal development of muscle tissue. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
| Accession Number | P51114 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 8087 |
| Name | Human FXR1 (aa 552-620) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1110050J02Rik; 9530073J07Rik; AI851072; FMR1 autosomal homolog 1; fragile x mental retardation autosomal homolog variant p1K; fragile x mental retardation autosomal homolog variant p2K; fragile x mental retardation autosomal homolog variant p4K; fragile x mental retardation autosomal homolog variant p5FK; fragile x mental retardation gene 1, autosomal homolog; fragile x mental retardation gene, autosomal homolog; fragile x mental retardation syndrome-related protein 1; fragile x mental retardation, autosomal homolog 1; fragile x mental retardation-related protein 1; fragile-X-related protein 1; Fxr1; Fxr1h; Fxr1p; hFXR1p; hypothetical protein LOC536793; mFxr1p |
| Common Name | FXR1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction