missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FXR/NR1H4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3554-SP
This item is not returnable.
View return policy
Description
FXR/NR1H4 Polyclonal specifically detects FXR/NR1H4 in Human samples. It is validated for Western Blot.
Specifications
| FXR/NR1H4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BAR, bile acid receptor, farnesoid X receptor, Farnesoid X-activated receptor, Farnesol receptor HRR-1, FXRBAR, HRR-1, HRR1MGC163445, NR1H4, Nuclear receptor subfamily 1 group H member 4, nuclear receptor subfamily 1, group H, member 4, Retinoid X receptor-interacting protein 14, RIP14, RIP14RXR-interacting protein 14 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9971 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q96RI1 | |
| NR1H4 | |
| E. coli-derived recombinant human FXR/NR1H4 Leu211-Gln486 Accession # Q96RI1 | |
| 25 μg | |
| Primary | |
| Detects human FXR/NR1H4 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction