missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Furin Recombinant Protein, PeproTech®

Product Code. 30088443
Click to view available options
Quantity:
2 x 50 μg
5 x 50 μg
Unit Size:
Pack of 2
Pack of 5
This item is not returnable. View return policy

Product Code. 30088443

Brand: Gibco™ 45047100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Furin is a membrane-associated, calcium-dependent, serine protease that belongs to the subtilisin-like prohormone convertase (PC) family. Members of this family of cellular enzymes cleave most prohormones and neuropeptide precursors. Numerous other cellular proteins, some viral proteins, and bacterial toxins that are transported by the constitutive secretory pathway are also targeted for maturation by PCs. Furin and other PC family members share structural similarities which include a heterogeneous ~10 kDa amino-terminal proregion, a highly conserved ~55 kDa subtilisin-like catalytic domain, and carboxyl-terminal domain that is heterogeneous in length and sequence. These enzymes become catalytically active following proregion cleavage within the appropriate cellular compartment. Furin is the only known PC to possess a transmembrane domain. Cleavage of target proteins occurs at the carboxyl-terminus of the furin consensus sequence, RX(K/R)R. It has been shown that the acidic peptide sequence, C771PSDSEEDEG780, localizes furin to the trans-Golgi-network. Phosphorylation of serine residues within this region modulates intracellular routing of furin protein. An additional signaling domain includes the tetrapeptide sequence, Y759KGL762, which directs internalization from the cell surface.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09958
For Use With (Application) Functional Assay
Formulation Protein with no preservative
Gene ID (Entrez) 5045
Molecular Weight (g/mol) 63.9 kDa
Name Furin
Quantity 2 x 50 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias 9130404I01Rik; dibasic processing enzyme; dibasic-processing enzyme; FES upstream region; FUR; Furin; furin (paired basic amino acid cleaving enzyme); furin, membrane associated receptor protein; furin, paired basic amino acid cleaving enzyme; furin; LOW QUALITY PROTEIN: furin; Pace; Paired basic amino acid cleaving enzyme (furin); paired basic amino acid residue cleaving enzyme; paired basic amino acid residue-cleaving enzyme; Pcsk3; prohormone convertase 3; proprotein convertase subtilisin/kexin type 3; Proprotein convertase subtilisin/kexin type 3 (paired basic amino acid cleaving enzyme, furin, membrane associated receptor protein); proprotein convertase subtilisin/kexin type3; SPC1; subtilisin pro-protein processing enzyme
Gene Symbol FURIN
Conjugate Unconjugated
Recombinant Recombinant
Activity Measured by its ability to cleave the fluorogenic peptide substrate Boc-Arg-Val-Arg-Arg-AMC (Bachem Catalog# I-1645.0025).
Expression System Insect cells
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.