Learn More
Human Fucosyltransferase 8/FUT8 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand:  R&D Systems AF5768
Product Code. 18051762
  • £506.00 / 100µg
Description
Fucosyltransferase 8/FUT8 Polyclonal specifically detects Fucosyltransferase 8/FUT8 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fucosyltransferase 8/FUT8
Polyclonal
Unconjugated
Q9BYC5
FUT8
Chinese hamster ovary cell line CHO-derived recombinant human Fucosyltransferase 8/FUT8 Asp32-Lys575 Accession # Q9BYC5
100 μg
Primary
Detects human Fucosyltransferase 8/FUT8 in direct ELISAs and Western blots.
Human
IgG
Western Blot, Immunohistochemistry, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL
alpha 1,6 Fucosyltransferase, alpha-(1,6)-fucosyltransferase, alpha1-6FucT, EC 2.4.1.68, Fucosyltransferase 8, fucosyltransferase 8 (alpha (1,6) fucosyltransferase), FUT8, GDP-fucose--glycoprotein fucosyltransferase, GDP-L-Fuc:N-acetyl-beta-D-glucosaminide alpha1,6-fucosyltransferase, Glycoprotein 6-alpha-L-fucosyltransferase, MGC26465
Sheep
Affinity Purified
RUO
2530
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.