missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fucosyltransferase 11/FUT11 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5964
This item is not returnable.
View return policy
Description
Fucosyltransferase 11/FUT11 Polyclonal specifically detects Fucosyltransferase 11/FUT11 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunocytochemistry.
Specifications
| Fucosyltransferase 11/FUT11 | |
| Polyclonal | |
| Unconjugated | |
| AAH36037 | |
| FUT11 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Fucosyltransferase 11/FUT11 Gly25-Leu492 Accession # AAH36037 | |
| 100 μg | |
| Primary | |
| Detects human FUT11 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) FUT3, rhFUT5, and rhFUT8 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| alpha-(1,3)-fucosyltransferase 11, EC 2.4.1, EC 2.4.1.-, Fucosyltransferase 11, fucosyltransferase 11 (alpha (1,3) fucosyltransferase), Fucosyltransferase XI, Fuc-TXI, FucT-XI, FUT11, Galactoside 3-L-fucosyltransferase 11, MGC119338, MGC119339, MGC33202 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 170384 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction