missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human FRP-5 (soluble) Recombinant Protein, PeproTech®

Product Code. 17871023
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17871023

Brand: Gibco™ 12053250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

PeproTech's CHO cell-derived Recombinant Human sFRP5 is a 288-amino-acid length glycoprotein with a calculated molecular weight of 32.7 kDa; however, due to glycosylation, protein migration occurs at an apparent molecular weight of approximately 33-35 kDa by SDS-PAGE analysis under both reducing and non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Secreted frizzled-related protein 5 (SFRP5) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. SFRP5 and SFRP1 may be involved in determining the polarity of photoreceptor cells in the retina. SFRP5 is highly expressed in the retinal pigment epithelium, and moderately expressed in the pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5T4F7
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 6425
Molecular Weight (g/mol) 32.7 kDa
Name Human FRP-5 (soluble)
Quantity 250 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AI605071; Frizzled-related protein 1 b; FRP1B; FRP-1 b; SARP3; SARP-3; secreted apoptosis related protein 3; secreted apoptosis-related protein 3; secreted frizzled related protein 5; secreted frizzled-related protein 5; secreted frizzled-related sequence protein 5; SFRP5; sFRP-5
Common Name SFRP5
Gene Symbol SFRP5
Biological Activity Determined by its ability to decrease alkaline phosphatase activity in CCL-226 cells when treated with 25 ng/ml of Murine Wnt-3 a.
Conjugate Unconjugated
Recombinant Recombinant
Sequence EEYDYYGWQA EPLHGRSYSK PPQCLDIPAD LPLCHTVGYK RMRLPNLLEH ESLAEVKQQA SSWLPLLAKR CHSDTQVFLC SLFAPVCLDR PIYPCRSLCE AVRAGCAPLM EAYGFPWPEM LHCHKFPLDN DLCIAVQFGH LPATAPPVTK ICAQCEMEHS ADGLMEQMCS SDFVVKMRIK EIKIENGDRK LIGAQKKKKL LKPGPLKRKD TKRLVLHMKN GAGCPCPQLD SLAGSFLVMG RKVDGQLLLM AVYRWDKKNK EMKFAVKFMF SYPCSLYYPF FYGAAEPH
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.