missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Frizzled-10 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£176.00 - £467.00
Specifications
| Antigen | Frizzled-10 |
|---|---|
| Clone | 2678A |
| Dilution | Immunohistochemistry 0.3-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Immunohistochemistry, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18733447
![]() |
R&D Systems
MAB34581-SP |
25 μg |
£176.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18713797
![]() |
R&D Systems
MAB34581-100 |
100 μg |
£467.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Frizzled-10 Monoclonal antibody specifically detects Frizzled-10 in Human samples. It is validated for Immunohistochemistry, Flow CytometrySpecifications
| Frizzled-10 | |
| Immunohistochemistry 0.3-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD350, CD350 antigen, frizzled (Drosophila) homolog 10, frizzled homolog 10 (Drosophila), frizzled-10, Frizzled10, Fz10, Fz-10, FZD10, FzE7, hFz10 | |
| Mouse myeloma cell line NS0-derived human Frizzled-10, Ile21-Gly161, Accession # Q6NSL8 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2678A | |
| Immunohistochemistry, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q6NSL8 | |
| 11211 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title