missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Free BDNF Quantikine ELISA Kit
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
Description
The Human Free BDNF Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human BDNF in cell culture supernates (50 ul), serum (10 ul), edta plasma (50 ul), platelet-poor heparin plasma (50 ul), citrate plasma (50 ul).
Specifications
Specifications
| Assay Range | 62.50 - 4000pg/mL |
| Assay Sensitivity | 20pg/mL |
| Conjugate | HRP |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. Cross-reactivity observed with 1 or more available related molecules. |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate), Platelet-poor Plasma (Heparin), Serum |
| Specificity | Natural and Recombinant Human BDNF |
| Sufficient For | 1 x 96-well Plate |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction