missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FPRL2/FPR3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£168.00 - £369.00
Specifications
| Antigen | FPRL2/FPR3 |
|---|---|
| Clone | 374822 |
| Concentration | LYOPH |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18779397
![]() |
R&D Systems
MAB3896-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18101401
|
R&D Systems
MAB3896 |
100 μg |
£369.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FPRL2/FPR3 Monoclonal specifically detects FPRL2/FPR3 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| FPRL2/FPR3 | |
| LYOPH | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| P25089 | |
| 2359 | |
| HEK293 human embryonic kidney cell line transfected with human FPRL2 Met1-Met352 Accession # P25089 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human FPRL2. Stains human FPRL2 transfectants but not irrelevant transfectants. |
| 374822 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FMLP-R-II, FMLPY, formyl peptide receptor 3, Formyl peptide receptor-like 2FMLP-related receptor II, FPR3, FPRH1, FPRH1FPRH2, FPRH2, FPRL2FML2_HUMAN, N-formyl peptide receptor 3, RMLP-R-I | |
| FPR3 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title