missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FPR1 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems
Additional Details : Weight : 0.00970kg
Description
FPR1 Monoclonal specifically detects FPR1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
FPR1 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
NP_002020 | |
FPR1 | |
HEK293 human embryonic kidney cell line transfected with human FPR1 Met1-Lys350 Accession # NP_002020 | |
100 μg | |
Primary | |
Detects human FPR1. Stains human FPR1-transfected cells but not irrelevant transfectants. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
350418 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
fMet-Leu-Phe receptor, fMLF-R, FMLP, fMLP receptor, formyl peptide receptor 1, FPRfMet-Leu-Phe receptor, N-formyl peptide receptor, N-formylpeptide chemoattractant receptor | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2357 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
For Research Use Only