missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FoxP1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF4534
This item is not returnable.
View return policy
Description
FoxP1 Polyclonal specifically detects FoxP1 in Human samples. It is validated for Western Blot.
Specifications
| FoxP1 | |
| Polyclonal | |
| Unconjugated | |
| Q9H334 | |
| FOXP1 | |
| E. coli-derived recombinant human FoxP1 Leu231-Gly420 Accession # Q9H334 | |
| 100 μg | |
| Primary | |
| Detects human FoxP1 in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| FLJ23741, fork head-related protein like B, forkhead box P1, forkhead box protein P1, glutamine-rich factor 1, hFKH1B, HSPC215, MGC12942, MGC88572, MGC99551, QRF1, QRF112CC4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27086 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction