missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FOXO1 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1086
Code : NEW
Additional Details : Weight : 0.01000kg
Description
FOXO1 Synthetic Peptide, PEP-1086, from Invitrogen.
FOXO1 (FKHR, ForKHead Box 01) is a member in a subfamily of the forkhead homeotic gene family of transcription factors. Recent experiments have shown that FOXO1 can act as either a coactivator or a corepressor of nuclear receptor activity that is mediated through the LXXLL motif found in the carboxyl terminal region of the FKHR protein. Association of FOXO1 with PAX3 has been implicated in alveolar rhabdomyosarcoma. Recent studies link the anti-tumor activity of FOXO1, the process of autophagy and myogenic growth and differentiation. FOXO1 is the main target of insulin signaling that regulated the metabolic homeostasis in response to oxidative stress. FOXO1 binds to the insulin response element (IRE) with consensus sequence 5′-TT[G/A]TTTTG-3′ and the related Daf-16 family binding element (DBE) with consensus sequence 5′- TT[G/A]TTTAC-3′. FOXO1 is a regulator of redox balance, osteoblast numbers and controls bone mass. Further, FOXO1 orchestrates the endocrine function of the skeleton in regulating glucose metabolism, and suppresses the transcriptional activity of RUNX2, an upstream activator of osteocalcin/BGLAP. In hepatocytes, FOXO1 promotes gluconeogenesis by acting together with PPARGC1A to activate the expression of genes such as IGFBP1, G6PC and PPCK1. FOXO1 is an important regulator of cell death acting downstream of CDK1, PKB/AKT1 and SKT4/MST1. This peptide corresponds to 15 amino acids near the amino terminus of human FOXO1.PEP-1086 can be used as a blocking peptide with polyclonal antibody PA5-20972.Specifications
2308 | |
purified | |
Human FOXO1 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q12778 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
FOXO1 | |
0.02% sodium azide | |
50 μg | |
PBS |