missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FoxF2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6988-SP
This item is not returnable.
View return policy
Description
FoxF2 Polyclonal specifically detects FoxF2 in Human samples. It is validated for Western Blot.
Specifications
| FoxF2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FKHL6, FKHL6forkhead box protein F2, forkhead box F2, forkhead-like 6, Forkhead-related activator 2, Forkhead-related protein FKHL6, Forkhead-related transcription factor 2, FREAC2, FREAC2FREAC-2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 2295 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q12947 | |
| FOXF2 | |
| E. coli-derived recombinant human FoxF2 Ala324-Met444 Accession # Q12947 | |
| 25 μg | |
| Primary | |
| Detects human FoxF2 in direct ELISAs and Western blots. In direct ELISAs, less than 3% cross-reactivity with recombinant human (rh) FoxF1, rhFoxP3, and rhFoxC2 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction