missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FOXD4L1 Partial ORF (NP_036316.1, 285 a.a. - 384 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16172377
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16172377 25 μg 25µg
16162377 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16172377 Supplier Abnova™ Supplier No. H00200350Q01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

This gene is a member of the forkhead/winged-helix (FOX) family of transcription factors with highly conserved FOX DNA-binding domains. Members of the FOX family of transcription factors are regulators of embryogenesis and may play a role in human cancer. This gene lies in a region of chromosome 2 that surrounds the site where two ancestral chromosomes fused to form human chromosome 2. This region is duplicated elsewhere in the human genome, primarily in subtelomeric and pericentromeric locations, thus mutiple copies of this gene have been found. [provided by RefSeq]

Sequence: EGADLATPGTLPVLQPSLGPQPWEEGKGLASPPGGGCISFSIESIMQGVRGAGTGAAQSLSPTAWSYCPLLQRPSSLSDNFAATAAASGGGLRQRLRSHQ

Specifications

Accession Number NP_036316.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 200350
Molecular Weight (g/mol) 36.74kDa
Name FOXD4L1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen EGADLATPGTLPVLQPSLGPQPWEEGKGLASPPGGGCISFSIESIMQGVRGAGTGAAQSLSPTAWSYCPLLQRPSSLSDNFAATAAASGGGLRQRLRSHQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FOXD5/bA395L14.1
Common Name FOXD4L1
Gene Symbol FOXD4L1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.