missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FoxD3 aa 1-140 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF2819
This item is not returnable.
View return policy
Description
FoxD3 Polyclonal specifically detects FoxD3 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| FoxD3 | |
| Polyclonal | |
| Unconjugated | |
| Q9UJU5 | |
| FOXD3 | |
| E. coli-derived recombinant human FoxD3 Met1-Val140 Accession # Q9UJU5 | |
| 100 μg | |
| Primary | |
| Detects human FoxD3 (aa 1-140) in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) FoxP3, rhFoxP1, and rhFoxO3A is observed. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| AIS1, CWH3, forkhead box D3, forkhead box protein D3, Genesis, HFH2, HNF3/FH transcription factor genesis | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 27022 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction