missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FOXC1 (aa 497-553) Control Fragment Recombinant Protein

Product Code. 30181035
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30181035

Brand: Invitrogen™ RP98335

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

FOXC1 is a protein belonging to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this protein is not known, however, it plays a role in the regulation of FGF19-FGFR4-MAPK pathway to promote both the development and maintenance of anterior segment structures within the eye. Mutations in this gene cause various glaucoma, iridogoniodysgenesis anomaly, Peters anomaly that includes central corneal leukoma, absence of the posterior corneal stroma and Descemetmembrane and Axenfeld-Rieger anomaly characterized by posterior corneal embryotoxon, iris adhesion to the Schwalbe line, hypertelorism, hypoplasia of the malar bones, congenital absence of some teeth and mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q12948
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 2296
Name Human FOXC1 (aa 497-553) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias ARA; ch; congenital hydrocephalus; Fkh1; fkh-1; Fkhl7; forkhead box C1; forkhead box C1 protein; forkhead box protein C1; forkhead, drosophila, homolog-like 7; forkhead/winged helix-like transcription factor 7; forkhead-related activator 3; forkhead-related protein FKHL7; Forkhead-related transcription factor 3; Foxc1; FREAC3; FREAC-3; frkhda; HGNC:3800; IGDA; IHG1; IRID1; Mesoderm/mesenchyme forkhead 1; Mf1; MF-1; Mf4; myeloid factor-delta; RIEG3; Transcription factor FKH-1
Common Name FOXC1
Gene Symbol FOXC1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence YPGQQQNFHSVREMFESQRIGLNNSPVNGNSSCQMAFPSSQSLYRTSGAFVYDCSKF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.