missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Human, Forward Primer, HPLC

Product Code. 15594364 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
50 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15594364 50 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15594364 Supplier Applied Biosystems™ Supplier No. A15631
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Human, Forward Primer, HPLC

Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Format Solid
Description Human Forward HPLC Primer, 50 nmole, Solid, For PCR and Sanger Sequencing
Product Type Human Forward HPLC Primer
Primer Forward
Quantity 50 nmol
Grade HPLC
For Use With (Application) For PCR and Sanger Sequencing
Form Solid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.