missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Follistatin Recombinant Protein, PeproTech®

Product Code. 17831043
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17831043 5 μg 5µg
17841043 100 μg 100µg
17830173 1 mg 1mg
17820283 20 μg 20µg
17891513 250 μg 250µg
17831323 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17831043 Supplier Gibco™ Supplier No. 120135UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Follistatin is a 31.5 kDa protein containing 288 amino acids. Its primary structure contains three cysteine-rich domains (called FS domains), each followed by a protease-inhibitory kazal domain. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Follistatin is a single-chain gonadal protein that specifically inhibits follicle-stimulating hormone release. The single FST gene encodes two isoforms, FST317 and FST344 containing 317 and 344 amino acids respectively, resulting from alternative splicing of the precursor mRNA. In a study in which 37 candidate genes were tested for linkage and association with polycystic ovary syndrome (PCOS) or hyperandrogenemia in 150 families, evidence was found for linkage between PCOS and follistatin.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P19883
For Use With (Application) Control, Western Blot Control, ELISA Standard, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 10468
Molecular Weight (g/mol) 31.5 kDa
Name Human Follistatin
Purification Method Purified
Quantity 5 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias activin-binding protein; AL033346; FOL1; Follictatin; Follistatin; follistatin isoform FST317; follistatin-A; FS; FST; Fst-288; RATFOL1
Common Name Follistatin
Gene Symbol Fst
Biological Activity Determined by its ability to neutralize Activin A inhibitory effect of murine MPC-11 cells. The expected ED50 is 0.1-0.4 ug/ml in the presence of 7.5 ng/ml Activin A.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence GNCWLRQAKN GRCQVLYKTE LSKEECCSTG RLSTSWTEED VNDNTLFKWM IFNGGAPNCI PCKETCENVD CGPGKKCRMN KKNKPRCVCA PDCSNITWKG PVCGLDGKTY RNECALLKAR CKEQPELEVQ YQGRCKKTCR DVFCPGSSTC VVDQTNNAYC VTCNRICPEP ASSEQYLCGN DGVTYSSACH LRKATCLLGR SIGLAYEGKC IKAKSCEDIQ CTGGKKCLWD FKVGRGRCSL CDELCPDSKS DEPVCASDNA TYASECAMKE AACSSGVLLE VKHSGSCN
Content And Storage -20°C
Activity Determined by its ability to neutralize Activin A inhibitory effect of murine MPC-11 cells. The expected ED50 is 0.1-0.4 ug/ml in the presence of 7.5 ng/ml Activin A.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.