missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FNBP1 (aa 59-139) Control Fragment Recombinant Protein

Product Code. 30196806
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30196806

Brand: Invitrogen™ RP109139

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-139887 (PA5-139887. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a member of the formin-binding-protein family. The protein contains an N-terminal Fer/Cdc42-interacting protein 4 (CIP4) homology (FCH) domain followed by a coiled-coil domain, a proline-rich motif, a second coiled-coil domain, a Rho family protein-binding domain (RBD), and a C-terminal SH3 domain. This protein binds sorting nexin 2 (SNX2), tankyrase (TNKS), and dynamin; an interaction between this protein and formin has not been demonstrated yet in human.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96RU3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23048
Name Human FNBP1 (aa 59-139) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1110057E06Rik; 2210010H06Rik; FBP1; FBP17; Fnbp1; formin binding protein 1; formin binding protein 17; formin-binding protein 1; Formin-binding protein 17; hFBP17; Kiaa0554; MGC126804; Rapostlin
Common Name FNBP1
Gene Symbol FNBP1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SKEEEEYKYTSCKAFISNLNEMNDYAGQHEVISENMASQIIVDLARYVQELKQERKSNFHDGRKAQQHIETCWKQLESSKR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.