missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FMNL2 (aa 393-516) Control Fragment Recombinant Protein

Product Code. 30199250
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30199250

Brand: Invitrogen™ RP88596

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52148 (PA5-52148. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Formin-like protein 2 is a cytoplasmic phosphoprotein belonging to the formin homology domain containing protein family and has a DAD (diaphanous autoregulatory domain), FH2 (formin homology 2) domain, and GBD/FH3 (Rho GTPase-binding/formin homology 3) domain. The exact function of FMNL2 is yet to be known but reports suggest that it may have a role in the Wnt signaling pathway. Formin-related proteins are also known to play an important role in morphogenesis, cytokinesis and cell polarity. Several alternatively spliced transcript variants have been described but their full-length nature is not yet determined. It is also known to be associated with a diffuse-type gastric cancer, breast cancer, chondrosarcoma, melanoma, and glioblastoma. FMNL2 might be implicated in polarity control, invasion, migration or metastasis through actin filament nucleation and may be regulated by Rho family GTPase.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96PY5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 114793
Name Human FMNL2 (aa 393-516) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5430425K04Rik; FHOD2; FMNL2; formin homology 2 domain containing 2; formin homology 2 domain-containing protein 2; formin like 2; formin-like 2; formin-like domain containing protein MAN; formin-like protein 2; Kiaa1902; Man; protein Man
Common Name FMNL2
Gene Symbol FMNL2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ERVEELEENISHLSEKLQDTENEAMSKIVELEKQLMQRNKELDVVREIYKDANTQVHTLRKMVKEKEEAIQRQSTLEKKIHELEKQGTIKIQKKGDGDIAILPVVASGTLSMGSEVVAGNSVGP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.