missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FLT3 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
FLT3 Synthetic Peptide, PEP-1490, from Invitrogen.
FLT3 is a tyrosine protein kinase that is a member of the growth factor receptor tyrosine kinase family. The receptor is activated by binding of the fms-related tyrosine kinase 3 ligand to the extracellular domain, which induces homodimer formation in the plasma membrane leading to autophosphorylation of the receptor. The activated receptor kinase subsequently phosphorylates and activates multiple cytoplasmic effector molecules in pathways involved in apoptosis, proliferation, and differentiation of hematopoietic cells in bone marrow. Mutations that result in the constitutive activation of this receptor result in acute myeloid leukemia and acute lymphoblastic leukemia.
Specifications
Specifications
| Gene ID (Entrez) | 2322 |
| Species | Human |
| Purification Method | purified |
| Accession Number | P36888 |
| Common Name | Human FLT3 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction