missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Flt-3 Ligand (FLT3L) Recombinant Protein

Fms-Like Tyrosine Kinase 3 Ligand (FLT3 Ligand, FLT3LG) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC9413

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10688118

  • £5130.00 / Each
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Human Flt-3 Ligand (FLT3L) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

FLT3 Ligand is a cytokine involved in hematopoeisis.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human OCI-AML5 cells

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture
Sequential Chromatography
FLT-3 Ligand
Lyophilized Human Flt-3 Ligand should be stored at 2-8°C, preferably desiccated. Store reconstituted Human Flt-3 Ligand at ≤ –20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.
< 0.1 ng/μg
Human
Mammalian
Recombinant, Ligand
FLT-3 Ligand
Gibco™
35 kDa
1mg
Human
Carrier-Free
1 mg
Wet Ice
Growth Factors & Receptors
Lyophilized
Signal Transduction, Stem Cell Research, Oncology
>95% by SDS-PAGE, >95% by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.